KEY465199424 Key Organics / BIONET

methyl 4-[(3,5-dimethyl-1H-pyrazol-1-yl)methyl]benzoate

  • MDL number MFCD05256293
  • Cas Number : 957010-45-6
  • Molecular Weight : 244.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88130

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/7X-0822_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart