KEY465199439 Key Organics / BIONET

5-(4-fluorobenzoyl)-2-(4-fluorophenyl)pyridine

  • MDL number MFCD05256308
  • Cas Number : 866135-75-3
  • Molecular Weight : 295.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88174

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/7X-0879_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart