KEY465199543 Key Organics / BIONET

5-benzoyl-N-(3,4-dichlorophenyl)-1,3-thiazol-2-amine

  • MDL number MFCD00139570
  • Cas Number : 339023-05-1
  • Molecular Weight : 349.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88512

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/8D-103_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart