KEY465199546 Key Organics / BIONET

3-acetyl-4-(4-methylbenzenesulfonyl)-2,5-dihydro-1lambda6-thiophene-1,1-dione

  • MDL number MFCD00139213
  • Cas Number : 339023-21-1
  • Molecular Weight : 314.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88519

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/8D-114_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart