KEY465199551 Key Organics / BIONET

2-{[4-(3,4-dichlorophenyl)piperazin-1-yl](dimethylamino)methylidene}propanedinitrile

  • MDL number MFCD00139237
  • Cas Number : 339023-43-7
  • Molecular Weight : 350.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88532

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/8D-135_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart