KEY465199632 Key Organics / BIONET

ethyl 4-chloro-1-(3,5-dichlorophenyl)-6-oxo-1,6-dihydropyridazine-3-carboxylate

  • MDL number MFCD00243739
  • Cas Number : 339030-94-3
  • Molecular Weight : 347.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88931

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/8G-349S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart