KEY465199642 Key Organics / BIONET

11,13-dimethoxy-3-methyl-4,8-dioxa-3-azatricyclo[7.4.0.0^{2,6}]trideca-1(13),9,11-triene

  • MDL number MFCD00232822
  • Cas Number : 400088-00-8
  • Molecular Weight : 251.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/89037

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/8G-951_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart