KEY465199683 Key Organics / BIONET

4-{2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]propanoyl}-N-(prop-2-en-1-yl)-1H-pyrrole-2-carboxamide

  • MDL number MFCD00794610
  • Cas Number : 339099-53-5
  • Molecular Weight : 385.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/89251

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/8J-020_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart