KEY465199747 Key Organics / BIONET

4-methyl-N,N-bis(prop-2-en-1-yl)benzene-1-sulfonamide

  • MDL number MFCD00226469
  • Cas Number : 50487-72-4
  • Molecular Weight : 251.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/89553

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8K-911_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart