KEY465199783 Key Organics / BIONET

(4E)-4-{2-[4-chloro-3-(trifluoromethyl)phenyl]hydrazin-1-ylidene}-3,4-dihydro-1H-2-benzopyran-1,3-dione

  • MDL number MFCD00665107
  • Cas Number : 339100-72-0
  • Molecular Weight : 368.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/89686

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/8L-738_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart