KEY465199853 Key Organics / BIONET

4-(4-chlorophenyl)-2-(3,4-dichlorophenoxy)pyridine-3-carbonitrile

  • MDL number MFCD01444230
  • Cas Number : 339103-64-9
  • Molecular Weight : 375.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/89868

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/8M-547S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart