KEY465199870 Key Organics / BIONET

(2E)-3-(furan-2-yl)-2-(4-phenyl-1,3-thiazol-2-yl)prop-2-enenitrile

  • MDL number MFCD00807578
  • Cas Number : 277308-31-3
  • Molecular Weight : 278.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/89908

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/8M-609S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart