KEY465199871 Key Organics / BIONET

(2E)-2-[4-(4-chlorophenyl)-1,3-thiazol-2-yl]-3-(3,4-dimethoxyphenyl)prop-2-enenitrile

  • MDL number MFCD01315155
  • Cas Number : 326915-13-3
  • Molecular Weight : 382.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/89909

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/8M-610S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart