KEY465199891 Key Organics / BIONET

5H,6H,7H,8H-imidazo[1,5-c]pyrimidin-5-one

  • MDL number MFCD01443896
  • Cas Number : 14509-66-1
  • Molecular Weight : 137.142

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/89956

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8M-743_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart