KEY465199929 Key Organics / BIONET

N2-tert-butyl-5-(4-fluorobenzoyl)-1,3-thiazole-2,4-diamine

  • MDL number MFCD01443916
  • Cas Number : 339106-68-2
  • Molecular Weight : 293.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90146

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/8N-720_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart