KEY465199963 Key Organics / BIONET

4-amino-5-benzoyl-2-{[(3-chlorophenyl)methyl]sulfanyl}thiophene-3-carbonitrile

  • MDL number MFCD02082264
  • Cas Number : 478081-59-3
  • Molecular Weight : 384.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90272

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/8P-391S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart