KEY465199966 Key Organics / BIONET

4-amino-5-benzoyl-2-{[2-(4-fluorophenyl)-2-oxoethyl]sulfanyl}thiophene-3-carbonitrile

  • MDL number MFCD02082268
  • Cas Number : 478081-63-9
  • Molecular Weight : 396.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90276

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/8P-395S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart