KEY465200000 Key Organics / BIONET

(Z)-({3-[(2-chloro-6-fluorophenyl)methyl]-2,4-dihydroxyphenyl}methylidene)(methyl)oxidoazanium

  • MDL number MFCD02082426
  • Cas Number : 478257-95-3
  • Molecular Weight : 309.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90358

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/8P-729_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart