KEY465200101 Key Organics / BIONET

N-(butan-2-yl)-4-propanoyl-1H-pyrrole-2-carboxamide

  • MDL number MFCD02571063
  • Cas Number : 478259-52-8
  • Molecular Weight : 222.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90558

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/8R-0279_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart