KEY465200151 Key Organics / BIONET

ethyl 2-[4-(4-fluorobenzoyl)-3,4-dihydro-2H-1,4-benzoxazin-3-yl]acetate

  • MDL number MFCD03012338
  • Cas Number : 439112-17-1
  • Molecular Weight : 343.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90648

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/8R-0398_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart