KEY465200170 Key Organics / BIONET

4-[2-(pyridin-3-yl)-1,3-thiazol-5-yl]pyrimidin-2-amine

  • MDL number MFCD28347964
  • Cas Number : 1823183-22-7
  • Molecular Weight : 255.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 263 - 265
Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/453115

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/8R-0803_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart