KEY465200190 Key Organics / BIONET

N-[2-(4-methanesulfonylpiperazin-1-yl)phenyl]-3-methylbenzamide

  • MDL number MFCD02571621
  • Cas Number : 478260-32-1
  • Molecular Weight : 373.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90751

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/8R-1027_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart