KEY465200200 Key Organics / BIONET

(2Z)-2-[(pyridin-2-yl)methylidene]-1-azabicyclo[2.2.2]octan-3-ol

  • MDL number MFCD11845414
  • Cas Number : 866136-79-0
  • Molecular Weight : 216.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90770

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/8R-1074_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart