KEY465200213 Key Organics / BIONET

{1,4,6-trimethyl-1H-pyrazolo[3,4-b]pyridin-3-yl}thiourea

  • MDL number MFCD27924225
  • Cas Number : 1797349-62-2
  • Molecular Weight : 235.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 269 - 271
Purity 97
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/402901

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/8T-0006_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart