KEY465200218 Key Organics / BIONET

1,3-dimethyl 2-[4-(2,5-dimethyl-1H-pyrrol-1-yl)phenyl]propanedioate

  • MDL number MFCD03618067
  • Cas Number : 866137-11-3
  • Molecular Weight : 301.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90893

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/8T-0093_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart