KEY465200236 Key Organics / BIONET

3-cyclopropyl-1-(2,5-dichlorobenzenesulfonyl)-1H-pyrazole

  • MDL number MFCD03305844
  • Cas Number : 956371-55-4
  • Molecular Weight : 317.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90945

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/8T-0280_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart