KEY465200239 Key Organics / BIONET

4-[(3-oxo-3,4-dihydro-2H-1,4-benzoxazin-6-yl)carbamoyl]butanoic acid

  • MDL number MFCD03305847
  • Cas Number : 866137-37-3
  • Molecular Weight : 278.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90950

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/8T-0285_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart