KEY465200257 Key Organics / BIONET

(2Z)-1-(4-chlorophenyl)-2-(3,5-dimethyl-1H-pyrazol-1-yl)-3-(dimethylamino)prop-2-en-1-one

  • MDL number MFCD03617312
  • Cas Number : 956792-44-2
  • Molecular Weight : 303.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90996

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/8T-0825_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart