KEY465200276 Key Organics / BIONET

4-benzyl-2-[4-(chlorodifluoromethyl)-6-methylpyrimidin-2-yl]-5-methyl-2,3-dihydro-1H-pyrazol-3-one

  • MDL number MFCD05668870
  • Cas Number : 866137-92-0
  • Molecular Weight : 364.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91070

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/8W-0307_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart