KEY465200299 Key Organics / BIONET

N-{4-[(4-oxo-2,3,4,5-tetrahydro-1H-1,5-benzodiazepin-1-yl)sulfonyl]phenyl}acetamide

  • MDL number MFCD04126170
  • Cas Number : 866138-30-9
  • Molecular Weight : 359.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91132

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/8W-0860_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart