KEY465200313 Key Organics / BIONET

(2E)-2-[(4-methoxyphenyl)methylidene]-2,3,4,9-tetrahydro-1H-carbazol-1-one

  • MDL number MFCD04126188
  • Cas Number : 1147118-58-8
  • Molecular Weight : 303.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91158

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/8W-0896_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart