KEY465200318 Key Organics / BIONET

8-(2-bromophenyl)-5,12,14-trioxa-2-azatetracyclo[7.7.0.0^{3,7}.0^{11,15}]hexadeca-1(16),3(7),9,11(15)-tetraen-6-one

  • MDL number MFCD05256348
  • Cas Number : 866138-52-5
  • Molecular Weight : 386.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91170

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/8X-0047_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart