KEY465200356 Key Organics / BIONET

N-[(E)-2-(1,3-dimethyl-4-nitro-1H-pyrazol-5-yl)ethenyl]aniline

  • MDL number MFCD05256399
  • Cas Number : 956626-42-9
  • Molecular Weight : 258.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91316

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/8X-0958_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart