KEY465200361 Key Organics / BIONET

7-bromo-2-(3-fluoro-4-hydroxyphenyl)-1,3-benzoxazol-5-ol

  • MDL number MFCD22375515
  • Cas Number : 544704-73-6
  • Molecular Weight : 324.105

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 261 - 265
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91339

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8Y-0027_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart