KEY465200393 Key Organics / BIONET

phenyl 3-(4-methoxyphenyl)-4-oxo-3,4-dihydrophthalazine-1-carboxylate

  • MDL number MFCD00138631
  • Cas Number : 338975-88-5
  • Molecular Weight : 372.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91494

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/9C-027_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart