KEY465200399 Key Organics / BIONET

4,6-dimethyl-2-({[4-(trifluoromethoxy)phenyl]carbamoyl}amino)pyridine-3-carboxamide

  • MDL number MFCD00138569
  • Cas Number : 338975-98-7
  • Molecular Weight : 368.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91506

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/9C-056_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart