KEY465200412 Key Organics / BIONET

methyl 2-({5-[(1Z)-(methoxyimino)methyl]imidazo[2,1-b][1,3]thiazol-6-yl}sulfanyl)benzoate

  • MDL number MFCD00139177
  • Cas Number : 338976-20-8
  • Molecular Weight : 347.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91547

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/9D-011_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart