KEY465200420 Key Organics / BIONET

6-(benzylsulfanyl)imidazo[2,1-b][1,3]thiazole-5-carbaldehyde

  • MDL number MFCD00139179
  • Cas Number : 338976-34-4
  • Molecular Weight : 274.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91569

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/9D-055_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart