KEY465200421 Key Organics / BIONET

3-[(2,4-dichlorophenyl)methyl]-6-hydrazinylpyridazine

  • MDL number MFCD00215384
  • Cas Number : 338976-38-8
  • Molecular Weight : 269.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91573

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/9D-060_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart