KEY465200424 Key Organics / BIONET

(5Z)-4-methyl-3-(4-nitrophenyl)-5-[(phenylamino)methylidene]-2,5-dihydrofuran-2-one

  • MDL number MFCD00139246
  • Cas Number : 338976-55-9
  • Molecular Weight : 322.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91595

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/9D-092_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart