KEY465200456 Key Organics / BIONET

1-(3-chlorophenyl)-4-[3-(trifluoromethyl)phenyl]-1H-pyrazol-5-amine

  • MDL number MFCD00140300
  • Cas Number : 324008-97-1
  • Molecular Weight : 337.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91700

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/9E-341S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart