KEY465200480 Key Organics / BIONET

2-[(6-chloro-2-phenyl-1H-1,3-benzodiazol-1-yl)oxy]acetic acid

  • MDL number MFCD00141352
  • Cas Number : 338978-62-4
  • Molecular Weight : 302.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91835

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/9F-324S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart