KEY465200485 Key Organics / BIONET

1,2-diphenyl-4,5,6,7-tetrahydro-1H-indol-4-one

  • MDL number MFCD00232414
  • Cas Number : 14073-27-9
  • Molecular Weight : 287.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91885

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/9F-906_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart