KEY465200503 Key Organics / BIONET

1-(5-nitropyridin-2-yl)-4-[3-(trifluoromethyl)phenyl]piperazine

  • MDL number MFCD01936536
  • Cas Number : 400088-85-9
  • Molecular Weight : 352.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91986

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/9G-335S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart