KEY465200554 Key Organics / BIONET

(2E)-3-(dimethylamino)-2-methanesulfonylprop-2-enenitrile

  • MDL number MFCD00202578
  • Cas Number : 858135-91-8
  • Molecular Weight : 174.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/92241

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9H-365S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart