KEY465200593 Key Organics / BIONET

6-chloro-2,4-bis(dimethylamino)pyridine-3-carbonitrile

  • MDL number MFCD00052641
  • Cas Number : 35022-97-0
  • Molecular Weight : 224.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/92409

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/9J-049_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart