KEY465200655 Key Organics / BIONET

ethyl 4-(2,6-dichlorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydropyridine-3-carboxylate

  • MDL number MFCD04126263
  • Cas Number : 866143-59-1
  • Molecular Weight : 328.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/92694

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/9L-515S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart