KEY465201205 Key Organics / BIONET

methyl 3-[4-(4-fluorobenzoyl)-1H-1,2,3-triazol-1-yl]thiophene-2-carboxylate

  • MDL number MFCD03012411
  • Cas Number : 439121-25-2
  • Molecular Weight : 331.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93741

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/9R-0807_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart