KEY465201213 Key Organics / BIONET

2-[1-(2-cyanophenyl)-2,5-dimethyl-1H-pyrrol-3-yl]-2-oxo-N-[4-(propan-2-yl)phenyl]acetamide

  • MDL number MFCD04126296
  • Cas Number : 866144-07-2
  • Molecular Weight : 385.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93758

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/9R-1018_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart