KEY465201312 Key Organics / BIONET

2-amino-3-[(E)-[(2,5-dimethoxyphenyl)methylidene]amino]-6-(trifluoromethyl)-3,4-dihydropyrimidin-4-one

  • MDL number MFCD04126356
  • Cas Number : 866145-76-8
  • Molecular Weight : 342.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94061

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/9W-0298_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart